Learn More
6-(2-(4-Methoxyphenyl)-1H-benzo[d]imidazol-5-yl)-5-methyl-4,5-dihydropyridazin-3(2H)-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 74150-27-9 |
| Molecular Formula | C19H18N4O2 |
| MDL Number | MFCD00761648 |
| Quantity | 1 g |
| SMILES | O=C1CC(C)C(C2=CC=C3C(N=C(C4=CC=C(OC)C=C4)N3)=C2)=NN1 |
| Molecular Weight (g/mol) | 334.37 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-(2-(4-Methoxyphenyl)-1H-benzo[d]imidazol-5-yl)-5-methyl-4,5-dihydropyridazin-3(2H)-one |