Learn More
PF 06650833
Click to view available options
:
5mg
Description
PF 06650833 is a potent and selective IRAK4 inhibitor (IC50 = 2 nM). Inhibits LPS-induced TNF-α secretion in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 1-[[(2S,3S,4S)-3-Ethyl-4-fluoro-5-oxo-2-pyrrolidinyl]methoxy]-7-methoxy-6-isoquinolinecarboxamide |
| CAS | 1817626-54-2 |
| Assay Percent Range | >98% |
| Quantity | 5 mg |
| Molecular Formula | C18H20FN3O4 |
| Formula Weight | Observed MW: 361.37 |