missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress ortho-iodoHoechst 33258

Product Code. 16449257
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449257 5 mg 5mg
16492777 50 mg 50mg
16476698 10 mg 10mg
16406856 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16449257 Supplier MedChemExpress Supplier No. HY15626
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ortho-iodoHoechst 33258 is part of a family of blue fluorescent dyes used to stain DNA.

Specifications

Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantity 5 mg
Shipping Condition Room temperature
Solubility DMSO : 50 mg/mL (93.56 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Molecular Weight (g/mol) 534.39 g/mol
CAS 158013-41-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.