Learn More
5beta-Pregnan-3alpha-ol-20-one-d4 (contains 10% toluene), TRC
Product Code. 30425178
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425178 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30425178 Supplier Toronto Research Chemicals Supplier No. TRCP7113021MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5beta-Pregnan-3alpha-ol-20-one-d4
CAS 96737-93-8
Molecular Formula C21 D4 H30 O2
Quantity 1 mg
Synonym Pregnan-20-one-17,21,21,21-d4','3-hydroxy-','(3α,5β)- (9CI), (3α,5β)-3-Hydroxypregnan-20-one-17,21,21,21-d4, 3α-Hydroxy-5β-pregnan-20-one-17,21,21,21-D4, Pregnanolone-17,21,21,21-d4, 3α,5β-Pregnanolone-17,21,21,21-d4, 3α,5β-Tetrahydroprogesterone-17,21,21,21-d4, 5β-Pregnan-3α-ol-20-one-17α,21,21,21-d4, 5β-Pregnan-3α-ol-20-one-d4
InChI Formula InChI=1S/C21H34O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h14-19,23H,4-12H2,1-3H3/t14-,15-,16+,17-,18+,19+,20+,21-/m1/s1/i1D3,17D
SMILES [2H]C([2H])([2H])C(=O)[C@@]1([2H])CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name 2,2,2-trideuterio-1-[(3R,5R,8R,9S,10S,13S,14S,17S)-17-deuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]ethanone
Molecular Weight (g/mol) 322.518
Formula Weight 322.281