Learn More
Fisherbrand™ Q55 Sonicator without Probe
Product Code. 12901161 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Voltage:
110 V
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Voltage Unit Size
12901161 110 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12901161
  • ONLINE EXCLUSIVE PRICE
    1970.00 EUR
    1588.04 EUR / Pack of 1
    Save 381.96 EUR
    19% Off
View all promotions
Estimated Shipment: 20-01-2027
Effective for standard cell disruption and many other small-volume applications. Fisherbrand™ Q55 Sonicator is a compact and cost-effective ultrasonic processor that will occupy less bench space than any unit on the market.
  • Smallest unit available
  • Thumb-switch or continuous operation
  • Simple and effective operation
Specifications
Voltage 110 V
Electrical Requirements 110V, 50/60 Hz
Wattage 55 W
Dimensions (L x W x H) 203 x 190 x 146 mm (8 x 7.5 x 5.75 in.)
Includes Generator, converter, 3 mm diameter probe, power cable, converter cable and wrench set
Model Q55