Learn More
DDX5 Antibody (2C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26627-50ul
 View more versions of this product
Product Code. 30043954
  • 343.00 EUR / 50µL
Description
DDX5 Monoclonal antibody specifically detects DDX5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DDX5
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
1655
Store at -20 to -70C. Avoid freeze-thaw cycles.
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
2C3
Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200
B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C
A synthesized peptide derived from Human DDX5 [UniProt P17844]
50 μL
Primary
Human, Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions