missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DDX5 Antibody (2C3), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 EUR - 533.00 EUR
Specifications
| Antigen | DDX5 |
|---|---|
| Clone | 2C3 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043954
|
Novus Biologicals
NBP3-26627-50ul |
50 μL |
343.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043257
|
Novus Biologicals
NBP3-26627-100ul |
100 μL |
533.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DDX5 Monoclonal antibody specifically detects DDX5 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, ImmunocytochemistrySpecifications
| DDX5 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 1655 | |
| IgG | |
| Affinity purified |
| 2C3 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| B-cell CLL/lymphoma 7 protein family member C, B-cell CLL/lymphoma 7C | |
| A synthesized peptide derived from Human DDX5 [UniProt P17844] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title