missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylphenol-d11, TRC

Product Code. 30179537
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179537 50 mg 50mg
30179003 10 mg 10mg
30179407 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179537 Supplier Toronto Research Chemicals Supplier No. TRCT80844650MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 362049-45-4
Molecular Formula C9 D11 H O
Molecular Weight (g/mol) 147.26
Synonym Phen-3,5-d2-ol, 2,4,6-tri(methyl-d3)- (9CI)
IUPAC Name 3,5-dideuterio-2,4,6-tris(trideuteriomethyl)phenol
SMILES [2H]c1c(c([2H])c(c(O)c1C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C9H12O/c1-6-4-7(2)9(10)8(3)5-6/h4-5,10H,1-3H3/i1D3,2D3,3D3,4D,5D
Formula Weight 147.1579
Chemical Name or Material 2,4,6-Trimethylphenol-d11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.