Learn More
Spectrum™ Labs Spectra/Por™ 3 3.5 kD MWCO Standard RC Dry Dialysis Kits
Click to view available options
Diameter (Metric):
11.5 mm
29 mm
34 mm
Length (Metric):
1.1 mL/cm vol./length
5 m
6.4 mL/cm vol./length
9.3 mL/cm vol./length
Quantity:
1 m Tubing/Kit
5 m Tubing/Kit
Unit Size:
1m
5m
Each
Description
Includes
- 1m or 5m of Spectra/Por 3 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water) or without glycerin (no soaking required)
Specifications
Specifications
| MWCO | 3.5 kDa |
| Membrane | Regenerated Cellulose |
| Diameter (Metric) | 11.5 mm |
| Length (Metric) | 1.1 mL/cm vol./length |
| Kit Contents | 5m roll of Spectra/Por™ 1, 2 or 3 dialysis tubing, 1 standard spectra/por closure (aids buoyancy), 1 weighted spectra/por closure (aids vertical orientation), 5 opening picks |
| Quantity | 5 m Tubing/Kit |
| For Use With (Application) | Ideal for small volume dialysis or evaluation of membrane for an application |