Learn More
5-Trifluoromethyl-2-thio-pyridone, TRC
Product Code. 30457330
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457330 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30457330 Supplier Toronto Research Chemicals Supplier No. TRCT7918651G
 Restricted Item
Estimated Shipment: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5-Trifluoromethyl-2-thio-pyridone
CAS 76041-72-0
Molecular Formula C6 H4 F3 N S
Quantity 1 g
InChI Formula InChI=1S/C6H4F3NS/c7-6(8,9)4-1-2-5(11)10-3-4/h1-3H,(H,10,11)
SMILES FC(F)(F)c1ccc(S)nc1
IUPAC Name 5-(trifluoromethyl)pyridine-2-thiol
Molecular Weight (g/mol) 179.16
Formula Weight 179.0017