Learn More
5-(Thiophen-2-yl)isoxazole-3-carboxylic Acid, TRC
Product Code. 30422606
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422606 500 mg -
30411222 1 g -
30411894 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30422606 Supplier Toronto Research Chemicals Supplier No. TRCT266791500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 763109-71-3
Molecular Formula C8H5NO3S
Molecular Weight (g/mol) 195.19
Synonym 5-(2-Thienyl)-3-isoxazolecarboxylic Acid
IUPAC Name 5-thiophen-2-yl-1,2-oxazole-3-carboxylic acid
SMILES OC(=O)c1cc(on1)c2cccs2
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C8H5NO3S/c10-8(11)5-4-6(12-9-5)7-2-1-3-13-7/h1-4H,(H,10,11)
Formula Weight 194.999
Chemical Name or Material 5-(Thiophen-2-yl)isoxazole-3-carboxylic Acid