missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 5-TAMRA, NHS
GREENER_CHOICE

Product Code. 18786526
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18786526

Brand: Tocris Bioscience™ 7665/1

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fluorescent amine-reactive dye for labeling proteins, peptides and oligonucleotides

5-TAMRA NHS is an orange-fluorescent dye that is commonly used for labeling proteins, peptides and oligonucleotides containing primary amines. Excitation maximum = 546 nm; emission maximum = 580 nm

Please see the protocol for further information on protein/antibody labeling and conjugation.

It is recommended to prepare stock solutions in DMSO.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[3-(Dimethylamino)-6-dimethylazaniumylidenexanthen-9-yl]-5-(2,5-dioxopyrrolidin-1-yl)oxycarbonylbenzoate
CAS 150810-68-7
Quantity 1 mg
Target Fluorescent Probes
Molecular Formula C29H25N3O7
Purity 0.9
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.