Learn More
(5-pyrid-4-ylthien-2-yl)methanol, 95%, Thermo Scientific™
Product Code. 10152834
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152834 1 g Amber Glass Bottle 1g
10081714 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10152834 Supplier Thermo Scientific Maybridge Supplier No. CC67009DA
 Restricted Item
Estimated Shipment: 04-01-2027
Chemical Identifiers
CAS 138194-04-4
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD09879963
InChI Key GDGUQNXUEMPEKM-UHFFFAOYSA-N
Synonym 5-pyrid-4-ylthien-2-yl methanol, 5-pyridin-4-yl thiophen-2-yl methanol, 5-4-pyridyl-2-thienyl methan-1-ol, 5-pyridin-4-ylthiophen-2-yl methanol
PubChem CID 15708918
IUPAC Name (5-pyridin-4-ylthiophen-2-yl)methanol
SMILES C1=CN=CC=C1C2=CC=C(S2)CO
Specifications
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 191.25
Percent Purity 95%
Chemical Name or Material (5-Pyrid-4-ylthien-2-yl)methanol
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.