missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Phenylisoxazole-3-carboxylic acid, ≥99%

Product Code. 15402829
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15402829 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15402829 Supplier Alfa Aesar™ Supplier No. H66772.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 14441-90-8
Melting Point 160°C to 164°C
Color White
Molecular Formula C10H7NO3
MDL Number MFCD00464221
Quantity 1 g
Beilstein 146162
Synonym 5-phenylisoxazole-3-carboxylic acid, 5-phenyl-isoxazole-3-carboxylic acid, 5-phenyl-3-isoxazolecarboxylic acid, 3-isoxazolecarboxylic acid, 5-phenyl, 3-isoxazolecarboxylicacid, 5-phenyl, 5-phenylisoxazole-3-carboxylicacid, 5-phenylisoxazol-3-carboxylic acid
InChI Key XJYOBHXWBRKOQO-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC(=NO2)C(=O)O
IUPAC Name 5-phenyl-1,2-oxazole-3-carboxylic acid
Molecular Weight (g/mol) 189.17
PubChem CID 151916
Formula Weight 189.17
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material 5-Phenylisoxazole-3-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.