missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1H-tetrazole, TRC

Product Code. 30422695
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30422695 500 mg -
30428375 250 mg -
30418268 2.5 g 2.5g
3 options
This item is not returnable. View return policy

Product Code. 30422695

Brand: Toronto Research Chemicals TRCP337023500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18039-42-4
Formula Weight 146.0592
InChI Formula InChI=1S/C7H6N4/c1-2-4-6(5-3-1)7-8-10-11-9-7/h1-5H,(H,8,9,10,11)
IUPAC Name 5-phenyl-1H-tetrazole
Molecular Formula C7 H6 N4
Molecular Weight (g/mol) 146.15
SMILES c1ccc(cc1)c2nnn[nH]2
Synonym 5-Phenyl-2H-tetrazole, 1H-Tetrazole, 5-phenyl- (8CI,9CI), Tetrazole, 5-phenyl- (6CI), 5-Phenyl-1,2,3,4-tetrazole, 5-Phenyl-1H-tetrazole, 5-Phenyl-2H-tetrazole, 5-Phenyltetrazole, Celltetra BHT-P 5T, Celltetra P 5T, Expandex 5PT, Expandex OX 5PT, Kempore 50XPT, MA 1623, NSC 11138, Phenyltetrazole, Unicell 5PT, Unifoam HB 1
Chemical Name or Material 5-Phenyl-1H-tetrazole

Specifications

Quantity 500 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.