accessibility menu, dialog, popup

UserName

CAS RN 18039-42-4

NNNN

CAS RN 18039-42-4

IUPAC Name: 5-phenyl-2H-1,2,3,4-tetrazol-2-ide
Synonyms: 5-phenyl-2H-1,2,3,4-tetrazol-2-ide
Molecular Weight (g/mol): 145.15
Molecular Formula: C7H5N4
InChi Key: LBCIMHAWIRLWHX-UHFFFAOYSA-N
SMILES: [N-]1N=NC(=N1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (8)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)