missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-1-pentyne, 98+%

Product Code. p-7045690
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392517 5 g 5g
11302527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392517 Supplier Thermo Scientific Alfa Aesar Supplier No. L11608.06
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1823-14-9 | C11H12 | 144.22 g/mol

5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Phenyl-1-pentyne is also used as an intermediate in organic synthesis and pharmaceuticals. It is also used to make other chemicals.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1823-14-9
Molecular Formula C11H12
Molecular Weight (g/mol) 144.22
MDL Number MFCD00039813
InChI Key KOSORCNALVBYBP-UHFFFAOYSA-N
Synonym 5-phenyl-1-pentyne, pent-4-yn-1-ylbenzene, benzene, 4-pentynyl, 5-phenylpentyne, unii-b4qv76z86q, pent-4-ynyl-benzene, ccris 7101, pent-4-yn-1-yl benzene, 5-phenyl pentyne, 4-pentynylbenzene #
PubChem CID 74573
IUPAC Name pent-4-ynylbenzene
SMILES C#CCCCC1=CC=CC=C1

Specifications

Density 0.95
Boiling Point 94°C (50 mmHg)
Flash Point 85°C (185°F)
Linear Formula C6H5(CH2)3C≡CH
Refractive Index 1.5165
Quantity 5 g
Beilstein 2074729
Formula Weight 144.22
Percent Purity ≥98%
Chemical Name or Material 5-Phenyl-1-pentyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.