Learn More
5’-O-Trityl Uridine, TRC
Product Code. 30455889
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455889 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455889 Supplier Toronto Research Chemicals Supplier No. TRCT890500250MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 5'-O-Trityl Uridine
CAS 6554-10-5
Molecular Formula C28 H26 N2 O6
Quantity 250 mg
Synonym Uridine','5'-O-(triphenylmethyl)-, Uridine','5'-O-trityl- (6CI,7CI,8CI), 5'-O-(Triphenylmethyl)uridine, 5'-O-Triphenylmethyluridine, 5'-O-Trityluridine, NSC 97038
InChI Formula InChI=1S/C28H26N2O6/c31-23-16-17-30(27(34)29-23)26-25(33)24(32)22(36-26)18-35-28(19-10-4-1-5-11-19,20-12-6-2-7-13-20)21-14-8-3-9-15-21/h1-17,22,24-26,32-33H,18H2,(H,29,31,34)/t22-,24-,25-,26-/m1/s1
SMILES O[C@@H]1[C@@H](COC(c2ccccc2)(c3ccccc3)c4ccccc4)O[C@H]([C@@H]1O)N5C=CC(=O)NC5=O
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(trityloxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 486.52
Formula Weight 486.1791