Learn More
5-Norbornene-2-carboxaldehyde, endo + exo, 95%
Product Code. 11481537
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11481537 25 g 25g
11491537 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11481537 Supplier Thermo Scientific Alfa Aesar Supplier No. B24973.14
 Restricted Item
In Stock
Add to basket
CAS: 5453-80-5 | C8H10O | 122.167 g/mol
5-Norbornene-2-carboxaldehyde was used in transition metal-catalyzed synthesis of norbornene-derived homopolymers having pendent t-Boc-protected quinizarin moieties for patterned fluorescence images. It acts as Diels-Alder adduct of acrolein and cyclopentadiene. It can be used as protected form of acrolein, from which the double bond may be liberated by cycloreversion, e.g. in a useful synthesis of 2-vinylimidazole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Norbornene-2-carboxaldehyde was used in transition metal-catalyzed synthesis of norbornene-derived homopolymers having pendent t-Boc-protected quinizarin moieties for patterned fluorescence images. It acts as Diels-Alder adduct of acrolein and cyclopentadiene. It can be used as protected form of acrolein, from which the double bond may be liberated by cycloreversion, e.g. in a useful synthesis of 2-vinylimidazole.
Solubility
Not miscible in water.
Notes
Store away from air and oxidizing agents. Airsensitive. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5453-80-5
Molecular Formula C8H10O
Molecular Weight (g/mol) 122.167
MDL Number MFCD00167568
InChI Key AJIBZRIAUXVGQJ-UHFFFAOYSA-N
Synonym 5-norbornene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carboxaldehyde, bicyclo 2.2.1 hept-5-ene-2-carbaldehyde, 2-formyl-5-norbornene, 5-formylbicyclohept-2-ene, 5-norbornene-2-carbaldehyde, 5-norbornane-2-carboxaldehyde, bicyclo 2.2.1 hept-2-ene-5-carbaldehyde
PubChem CID 95117
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carbaldehyde
SMILES C1C2CC(C1C=C2)C=O
Specifications
Density 1.032
Boiling Point 68°C to 70°C (12 mmHg)
Flash Point 50°C (122°F)
Odor Characteristic
Refractive Index 1.491
Quantity 25 g
UN Number UN1989
Beilstein 1363813
Sensitivity Air Sensitive
Solubility Information Not miscible in water.
Formula Weight 122.17
Percent Purity 95%
Chemical Name or Material 5-Norbornene-2-carboxaldehyde, endo + exo