missing translation for 'onlineSavingsMsg'
Learn More

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, MP Biomedicals™

Product Code. 11481151 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11481151 5 g 5g
11401161 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11481151 Supplier MP Biomedicals Supplier No. 0215775005
  • ONLINE EXCLUSIVE PRICE
    47.40 EUR
    45.15 EUR / 5g
    Save 2.25 EUR
    4% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, White Powder

  • It is useful as a nonionic density gradient medium.
  • It is a Non-ionic monomeric triiodinated x-ray contrast agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66108-95-0
Molecular Formula C19H26I3N3O9
Molecular Weight (g/mol) 821.14
MDL Number MFCD00077732
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
Synonym Nycodenz, Histodenz™

Specifications

Color White
Quantity 5 g
Beilstein 2406632
Physical Form Powder
Assay 0.99
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.