missing translation for 'onlineSavingsMsg'
Learn More

5-(N-2,3-Dihydroxypropylacetimido)-2,4,6-triiodo-N,N'-bis-(2,3-dihydroxypropyl)isophthalamide, 99.0%, MP Biomedicals™

Product Code. 11491151 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491151 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491151 Supplier MP Biomedicals Supplier No. 0215775025
  • ONLINE EXCLUSIVE PRICE
    128.00 EUR
    121.80 EUR / 25g
    Save 6.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 66108-95-0
Assay Percent Range 0.99
Molecular Formula C19H26I3N3O9
Quantity 25 g
Synonym Nycodenz
InChI Key NTHXOOBQLCIOLC-UHFFFAOYNA-N
SMILES CC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
IUPAC Name N1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide
Molecular Weight (g/mol) 821.14
Formula Weight 821.1
Physical Form Powder
Chemical Name or Material 5-(N-2, 3-Dihydroxypropylacetimido)-2, 4
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.