Learn More
5' Mod , Alexa Fluor 633 , 50 nmol
Description
Use this tool to design a custom DNA oligonucleotide, with any of a variety of 3' and 5' modifications, at scales ranging from 25 nmole to 10 μmole. Select desired purification method (normal or reversed-phase chromatography, HPLC, or PAGE).
Specifications
Specifications
| Quantity | 50 nmol |
| Product Line | Alexa Fluor |