missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-3-carbaldehyde, TRC

Product Code. 30335943
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335943 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30335943 Supplier Toronto Research Chemicals Supplier No. TRCB10135610MG
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Methylthiophene-3-carbaldehyde
CAS 29421-72-5
Molecular Formula C6 H6 OS
Quantity 10 mg
Synonym 4-Methyl-(9CI)-1H-imidazole-2-carboxylic Acid, 4-Methyl-1H-imidazole-2-carboxylic Acid, 4-Methylimidazole-2-carboxylic Acid
InChI Formula InChI=1S/C6H6OS/c1-5-2-6(3-7)4-8-5/h2-4H,1H3
SMILES Cc1cc(C=O)cs1
IUPAC Name 5-methylthiophene-3-carbaldehyde
Molecular Weight (g/mol) 126.176
Formula Weight 126.0139
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.