missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisophthalonitrile, 97%, Thermo Scientific™

Product Code. 15478959
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478959 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15478959 Supplier Thermo Scientific Alfa Aesar Supplier No. L12518.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39718-07-5
Melting Point 162°C to 166°C
Molecular Formula C9H6N2
MDL Number MFCD00052016
Quantity 1 g
UN Number UN3439
Beilstein 1940447
Synonym 3,5-dicyanotoluene, 5-methylisophthalonitrile, acmc-20and3, 5-methyl-isophthalonitrile, 1,3-benzenedicarbonitrile,5-methyl
InChI Key LYSGVAGOLJKEGK-UHFFFAOYSA-N
SMILES CC1=CC(=CC(=C1)C#N)C#N
IUPAC Name 5-methylbenzene-1,3-dicarbonitrile
Molecular Weight (g/mol) 142.161
PubChem CID 2736100
Formula Weight 142.16
Percent Purity 97%
Chemical Name or Material 5-Methylisophthalonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.