Learn More
MedChemExpress 5-Methylcytidine
Product Code. 16421037
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16421037 Supplier MedChemExpress Supplier No. HY113135
 Restricted Item
Estimated Shipment: 30-11-2026
MedChemExpress 5-Methylcytidine is a pyrimidine nucleoside detected in multiple biofluids.
Specifications
Molecular Weight (g/mol) 257.24 g/mol
Product Type Natural Products
Content And Storage -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Shipping Condition Room temperature
Purity 97.0%
Quantity 50 mg
Solubility Information DMSO : 130 mg/mL (505.36 mM∣Need ultrasonic)
For Use With (Application) Metabolism-protein/nucleotide metabolism
CAS 2140-61-6