Learn More
5-Methyl-1,3-oxazole-4-carboxylic acid, 95%, Thermo Scientific™
Product Code. 15294388 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15294388 5 g 5g
15284388 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15294388 Supplier Thermo Scientific Acros Supplier No. 454750050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 103879-58-9
Molecular Formula C5H5NO3
Molecular Weight (g/mol) 127.1
InChI Key QIACATCUODRSLS-UHFFFAOYSA-N
Synonym 5-methyloxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 5-methyl, 5-methyloxazole-4-carboxylic ac, 4-oxazolecarboxylicacid, 5-methyl, 5-methyl-oxazole-4-carboxylic acid, acmc-209zwx, 5-methyloxazole-4-carboxylicacid, 5-methyl-4-oxazolecarboxylic acid, 4-carboxy-5-methyl-1,3-oxazole, 5-methyl oxazole-4-carboxylic acid
PubChem CID 11355484
IUPAC Name 5-methyl-1,3-oxazole-4-carboxylic acid
SMILES CC1=C(N=CO1)C(=O)O
Specifications
Melting Point 149.0°C to 153.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 5 g
Formula Weight 127.1
Percent Purity 95%
Chemical Name or Material 5-Methyl-1,3-oxazole-4-carboxylic acid
compliance-icons
Product Identifier
  • 5-Methyl-1,3-oxazole-4-carboxylic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.