missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one, Mikromol™

Code produit. 30459716
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
100 mg
4 x 25 mg
missing translation for 'unitSize'
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity unitSize
30459716 4 x 25 mg -
30051930 100 mg 100mg
30051295 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 30459716 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0058.0400254AMP

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-Methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one, Mikromol
TRUSTED_SUSTAINABILITY

Spécification

Chemical Name or Material Fluvoxketone
CAS 61718-80-7
Analyte or Component Names Fluvoxketone
Shipping Condition Room Temperature
Molecular Formula C13 H15 F3 O2
Certifications/Compliance ISO 17025
Synonym Fluvoxamine Maleate Imp. D (EP), Fluvoxamine Imp. D (EP), 5-Methoxy-1-[4-(trifluoromethyl)phenyl]-1-pentanone, Fluvoxketone, Fluvoxamine Maleate Impurity D, Fluvoxamine Impurity D
InChI Formula InChI=1S/C13H15F3O2/c1-18-9-3-2-4-12(17)10-5-7-11(8-6-10)13(14,15)16/h5-8H,2-4,9H2,1H3
Solution Type Neat
SMILES COCCCCC(=O)c1ccc(cc1)C(F)(F)F
IUPAC Name 5-methoxy-1-[4-(trifluoromethyl)phenyl]pentan-1-one
Molecular Weight (g/mol) 260.25
Quantity 4 x 25 mg
Formula Weight 260.1024 g/mol
Afficher plus Afficher moins
Nom du produit
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.