missing translation for 'onlineSavingsMsg'
Learn More

5-Isopropyl-m-xylene (~85%), TRC

Product Code. 30406829
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406829 500 mg -
30402776 2.5 g 2.5g
30401559 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406829 Supplier Toronto Research Chemicals Supplier No. TRCI874653500MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4706-90-5
Molecular Formula C11H16
Molecular Weight (g/mol) 148.24
IUPAC Name 1,3-dimethyl-5-propan-2-ylbenzene
SMILES CC(C)c1cc(C)cc(C)c1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H16/c1-8(2)11-6-9(3)5-10(4)7-11/h5-8H,1-4H3
Formula Weight 148.1252
Chemical Name or Material 5-Isopropyl-m-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.