Learn More
5-Iodo-2-methylaniline, 98%, Thermo Scientific™
Product Code. 11371897
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11371897 5 g 5g
11361897 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11371897 Supplier Thermo Scientific Alfa Aesar Supplier No. L07404.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 83863-33-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00051531
InChI Key IOEHXNCBPIBDBZ-UHFFFAOYSA-N
Synonym 2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
PubChem CID 522802
IUPAC Name 5-iodo-2-methylaniline
SMILES CC1=CC=C(I)C=C1N
Specifications
Melting Point 46°C to 49°C
Quantity 5 g
UN Number UN2811
Beilstein 2078769
Sensitivity Light sensitive
Formula Weight 233.05
Percent Purity 98%
Chemical Name or Material 5-Iodo-2-methylaniline