missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2'-Deoxycytidine, MP Biomedicals™

Product Code. 11398899 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398899 5 g 5g
11388899 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398899 Supplier MP Biomedicals Supplier No. 0210206205
  • ONLINE EXCLUSIVE PRICE
    1520.00 EUR
    1444.80 EUR / 5g
    Save 75.20 EUR
    4% Off
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 611-53-0
Molecular Formula C9H12IN3O4
Molecular Weight (g/mol) 353.12
MDL Number MFCD00038063
InChI Key WEVJJMPVVFNAHZ-RRKCRQDMSA-N
Synonym 5-iodo-2'-deoxycytidine, ibacitabine, 2'-deoxy-5-iodocytidine, 4-amino-1-2r,4s,5r-4-hydroxy-5-hydroxymethyl tetrahydrofuran-2-yl-5-iodopyrimidin-2 1h-one, ibacitabine inn, ibacitabinum latin, ibacitabina spanish, cytidine, 2'-deoxy-5-iodo, iododeoxycytidine, unii-3ek8532dzv
PubChem CID 65050
IUPAC Name 4-amino-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodo-1,2-dihydropyrimidin-2-one
SMILES NC1=NC(=O)N(C=C1I)[C@H]1C[C@H](O)[C@@H](CO)O1

Specifications

Quantity 5 g
Formula Weight 353.1
Physical Form Solid
Chemical Name or Material 5-Iodo-2'-Deoxycytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.