missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytryptamine, MP Biomedicals™

Product Code. 11483152 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483152 1 g 1g
11443152 25 mg 25mg
11453152 50 mg 50mg
11463152 100 mg 100mg
11473152 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11483152 Supplier MP Biomedicals Supplier No. 0215376591
  • ONLINE EXCLUSIVE PRICE
    912.00 EUR
    868.35 EUR / 1g
    Save 43.65 EUR
    4% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 153-98-0
Molecular Formula C10H13ClN2O
Molecular Weight (g/mol) 212.677
InChI Key MDIGAZPGKJFIAH-UHFFFAOYSA-N
Synonym serotonin hydrochloride, 5-hydroxytryptamine hydrochloride, 3-2-aminoethyl-1h-indol-5-ol hydrochloride, serotonin hcl, 5-hydroxytryptamine hcl, unii-gkn429m9vs, ccris 4420, 1h-indol-5-ol, 3-2-aminoethyl-, monohydrochloride, gkn429m9vs, 3-2-aminoethyl indol-5-ol, chloride
PubChem CID 160436
IUPAC Name 3-(2-aminoethyl)-1H-indol-5-ol;hydrochloride
SMILES C1=CC2=C(C=C1O)C(=CN2)CCN.Cl

Specifications

Color Gray
Assay Percent Range 0.993
Quantity 1 g
Formula Weight 212.7
Physical Form Powder
Chemical Name or Material 5-Hydroxytryptamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.