Learn More
5-Hydroxymethyl-2-furancarbaldehyde, Sigma-Aldrich™
Product Code. 15896650
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896650 1 g 1g
15881111 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15896650 Supplier Sigma Aldrich Supplier No. 8206780001
 Restricted Item
Estimated Shipment: 26-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-47-0
Grade Research
Molecular Weight (g/mol) 126.11
Specifications
Melting Point 27°C to 30°C
Boiling Point 154°C to 155°C/16 hPa
MDL Number MFCD00003234
Quantity 1 g
Synonym 5-Hydroxymethyl-2-furancarbaldehyde, 5-(Hydroxymethyl)furfurol, 5-Hydroxymethyl-2-furancarbaldehyde