Learn More
5-(Hydroxymethyl)-1-(1-phenylpropan-2-yl)-1H-pyrrole-2-carbaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 5-(Hydroxymethyl)-1-(1-phenylpropan-2-yl)-1H-pyrrole-2-carbaldehyde |
| CAS | 2469014-63-7 |
| Molecular Formula | C15H17NO2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C15H17NO2/c1-12(9-13-5-3-2-4-6-13)16-14(10-17)7-8-15(16)11-18/h2-8,10,12,18H,9,11H2,1H3 |
| SMILES | CC(Cc1ccccc1)n2c(CO)ccc2C=O |
| Molecular Weight (g/mol) | 243.3 |