Learn More
5-Hydroxycytosine, TRC
Product Code. 30137454
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137454 10 g 10g
30012212 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137454 Supplier Toronto Research Chemicals Supplier No. TRCH90537510G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13484-95-2
Molecular Formula C4 H5 N3 O2
Molecular Weight (g/mol) 127.1
Synonym 2(1H)-Pyrimidinone, 6-amino-5-hydroxy-, 2(1H)-Pyrimidinone, 4-amino-5-hydroxy- (9CI), Cytosine, 5-hydroxy- (7CI,8CI), 6-Amino-5-hydroxy-2(1H)-pyrimidinone, 5-Hydroxycytosine
IUPAC Name 4-amino-5-hydroxy-1H-pyrimidin-2-one
SMILES NC1=NC(=O)NC=C1O
Specifications
Quantity 10 g
InChI Formula InChI=1S/C4H5N3O2/c5-3-2(8)1-6-4(9)7-3/h1,8H,(H3,5,6,7,9)
Formula Weight 127.0382
Chemical Name or Material 5-Hydroxycytosine