Learn More
5-Hydroxy-4-azaphenanthrene, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-4-azaphenanthrene |
| CAS | 33155-90-7 |
| Molecular Formula | C13H9NO |
| Quantity | 10 g |
| Synonym | Benzo[h]quinolin-10-ol, 10-Hydroxybenzo[h]quinoline |
| InChI Formula | InChI=1S/C13H9NO/c15-11-5-1-3-9-6-7-10-4-2-8-14-13(10)12(9)11/h1-8,15H |
| SMILES | Oc1cccc2ccc3cccnc3c12 |
| IUPAC Name | benzo[h]quinolin-10-ol |
| Molecular Weight (g/mol) | 195.22 |
| Formula Weight | 195.0684 |