Learn More
5-Hydroxy-2-methylindole, 98%, Thermo Scientific™
Product Code. 15417547
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 15417547
Brand: Thermo Scientific Alfa Aesar H35580.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13314-85-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00022714
InChI Key MDWJZBVEVLTXDE-UHFFFAOYSA-N
PubChem CID 83336
IUPAC Name 2-methyl-1H-indol-5-ol
SMILES CC1=CC2=CC(O)=CC=C2N1
Specifications
Melting Point 130°C to 132°C
Color Brown
Quantity 1 g
Sensitivity Light sensitive
Synonym 5-hydroxy-2-methylindole, 5-hydroxy-2-methyl indole, 1h-indol-5-ol, 2-methyl, 2-methyl-5-hydroxyindole, 2-methyl-5-indolol, pubchem8551, 5-hydroxy-2methylindole, 2-methyl-1h-indole-5-ol, 5-hydroxyl-2-methyl-indole, 1h-indol-5-ol,2-methyl
Formula Weight 147.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Hydroxy-2-methylindole