accessibility menu, dialog, popup

UserName

CAS RN 13314-85-7

H3CHNOH

CAS RN 13314-85-7

IUPAC Name: 2-methyl-1H-indol-5-ol
Synonyms: 2-methyl-1H-indol-5-ol
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: MDWJZBVEVLTXDE-UHFFFAOYSA-N
SMILES: CC1=CC2=CC(O)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results