Learn More
Justrite™ 5 Gallon Drain Can
Description
- Easy collection of used solvents
- Wide-mouth funnel catches drainage
- Built-in flame arrester
- Wide-mouth funnel catches drainage with no spill, no mess
- Removable funnel is plated steel
- Built-in flame arrester keeps fire from contents
- A 1.9 cm threaded plug allows safe disposal of waste liquids from the drain can into a permanent receptacle
- Optional drain can cover fits on top of funel to keep out dirt
Specifications
Specifications
| Capacity (Metric) | 5 Gallon |
| Capacity (English) | 757 L |
| Color | Yellow |
| Includes | Plated steel funnel |
| Material | Steel |