missing translation for 'onlineSavingsMsg'
Learn More

5-Fluorouracil, ∽99%, MP Biomedicals™

Product Code. p-4404371 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11408080 5 g 5g
11418080 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408080 Supplier MP Biomedicals Supplier No. 0210172205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 51-21-8
Molecular Formula C4H3FN2O2
Molecular Weight (g/mol) 130.08
MDL Number MFCD00006018
InChI Key GHASVSINZRGABV-UHFFFAOYSA-N
Synonym 5-fluorouracil, fluorouracil, 5-fu, fluoroplex, adrucil, efudex, carac, fluracil, fluoroblastin, kecimeton
PubChem CID 3385
ChEBI CHEBI:46345
IUPAC Name 5-fluoro-1,2,3,4-tetrahydropyrimidine-2,4-dione
SMILES FC1=CNC(=O)NC1=O

Specifications

Color White
Assay Percent Range ∽99%
Quantity 5 g
Formula Weight 130.08
Physical Form Powder
Chemical Name or Material 5-Fluorouracil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.