missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroorotic Acid, MP Biomedicals

Product Code. 11464551 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464551 1 g 1g
11444551 50 mg 50mg
11454551 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464551 Supplier MP Biomedicals Supplier No. 0215421091
  • ONLINE EXCLUSIVE PRICE
    1270.00 EUR
    1206.45 EUR / 1g
    Save 63.55 EUR
    5% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 703-95-7
Molecular Formula C5H3FN2O4
Molecular Weight (g/mol) 174.087
MDL Number MFCD00150658
InChI Key SEHFUALWMUWDKS-UHFFFAOYSA-N
Synonym 5-fluoroorotic acid, 5-fluoroorotate, fluoroorotic acid, 5-fluoro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid, orotic acid, 5-fluoro, 5-foa, 5-fluoro orotic acid, 5-fluorouracil-4-carboxylic acid, unii-7ia9ouc93e, 5-fluoroorotic acid van
PubChem CID 69711
ChEBI CHEBI:42535
IUPAC Name 5-fluoro-2,4-dioxo-1H-pyrimidine-6-carboxylic acid
SMILES C1(=C(NC(=O)NC1=O)C(=O)O)F

Specifications

Color Yellow
Assay Percent Range 98.3%
Quantity 1 g
Formula Weight 174.1
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroorotic Acid, Selective agent for yeast molecular genetics and uracil auxotrophs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.