missing translation for 'onlineSavingsMsg'
Learn More

5'-Fluoro-2'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 15404867
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404867 1 g 1g
15414867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404867 Supplier Thermo Scientific Alfa Aesar Supplier No. H32171.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 243863-39-0
Molecular Formula C9H6F4O
Molecular Weight (g/mol) 206.14
MDL Number MFCD00236274
InChI Key USFYRRQTKUSMNK-UHFFFAOYSA-N
Synonym 5'-fluoro-2'-trifluoromethyl acetophenone, 1-5-fluoro-2-trifluoromethyl phenyl ethanone, 5'-fluoro-2'-trifluoromethyl acetophenon, 1-acetyl-5-fluoro-2-trifluoromethyl benzene
PubChem CID 2774766
IUPAC Name 1-[5-fluoro-2-(trifluoromethyl)phenyl]ethanone
SMILES CC(=O)C1=C(C=CC(=C1)F)C(F)(F)F

Specifications

Refractive Index 1.4462
Quantity 1 g
Formula Weight 206.14
Percent Purity 97%
Chemical Name or Material 5'-Fluoro-2'-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.