Learn More
5-Desamino 5-Oxo-2,5-dihydro Lamotrigine, TRC
Product Code. 18050930
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050930 10 mg 10mg
18030060 50 mg 50mg
18051430 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18050930 Supplier Toronto Research Chemicals Supplier No. TRCD28847010MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 252186-78-0
Molecular Formula C9 H6 Cl2 N4 O
Molecular Weight (g/mol) 257.08
Synonym 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one, 1,2,4-Triazin-5(2H)-one, 3-amino-6-(2,3-dichlorophenyl)-, 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(2H)-one, 5-Desamino 5-Oxo-2,5-dihydro Lamotrigine
IUPAC Name 3-amino-6-(2,3-dichlorophenyl)-4H-1,2,4-triazin-5-one
SMILES NC1=NN=C(C(=O)N1)c2cccc(Cl)c2Cl
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C9H6Cl2N4O/c10-5-3-1-2-4(6(5)11)7-8(16)13-9(12)15-14-7/h1-3H,(H3,12,13,15,16)
Formula Weight 255.9919
Percent Purity >95%
Chemical Name or Material 3-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-5(4H)-one
Grade HPLC