Learn More
5'-Deoxy-5'-(methylthio)adenosine, TRC
Product Code. 30011031
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011031 25 mg 25mg
30008680 50 mg 50mg
30005846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30011031 Supplier Toronto Research Chemicals Supplier No. TRCD24260025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2457-80-9
Molecular Formula C11 H15 N5 O3 S
Molecular Weight (g/mol) 297.33
Synonym 5'-S-Methyl-5'-thioadenosine, 5'-(Methylthio)-5'-deoxyadenosine, 5'-(Methylthio)adenosine, 5'-Deoxy(methylthio)adenosine, 5'-Deoxy-5'-(methylthio)adenosine, 5'-S-Methylthioadenosine, NSC 335422, Vitamin L2, β-D-Ribofuranose, 1-(6-amino-9H-purin-9-yl)-1-deoxy-5-S-methyl-5-thio-
IUPAC Name (2R,3R,4S,5S)-2-(6-aminopurin-9-yl)-5-(methylsulfanylmethyl)oxolane-3,4-diol
SMILES CSC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)ncnc23
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C11H15N5O3S/c1-20-2-5-7(17)8(18)11(19-5)16-4-15-6-9(12)13-3-14-10(6)16/h3-5,7-8,11,17-18H,2H2,1H3,(H2,12,13,14)/t5-,7-,8-,11-/m1/s1
Formula Weight 297.0896
Percent Purity >95%
Chemical Name or Material 5'-Deoxy-5'-(methylthio)adenosine
Grade HPLC