Learn More
5-Cyanophthalide, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Cyanophthalide |
| CAS | 82104-74-3 |
| Molecular Formula | C9 H5 N O2 |
| Quantity | 5 g |
| Synonym | 1,3-Dihydro-1-oxo-5-isobenzofurancarbonitrile, 1-Oxo-1,3-dihydroisobenzofuran-5-carbonitrile, 5-Cyanophthalide |
| InChI Formula | InChI=1S/C9H5NO2/c10-4-6-1-2-8-7(3-6)5-12-9(8)11/h1-3H,5H2 |
| SMILES | O=C1OCc2cc(ccc12)C#N |
| IUPAC Name | 1-oxo-3H-2-benzofuran-5-carbonitrile |
| Molecular Weight (g/mol) | 159.14 |
| Formula Weight | 159.032 |