Learn More
5-Chloro Dasatinib, TRC
Product Code. 30138532
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138532 50 mg 50mg
30005500 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30138532 Supplier Toronto Research Chemicals Supplier No. TRCC36492550MG
 Restricted Item
Estimated Shipment: 08-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2468737-75-7
Molecular Formula C22H25Cl2N7O2S
Molecular Weight (g/mol) 522.45
Synonym N-(3,6-Dichloro-2-methylphenyl)-2-[[6-[4-(2-hydroxyethyl)-1-piperazinyl]-2-methyl-4-pyrimidinyl]amino]-5-thiazolecarboxamide;
IUPAC Name N-(3,6-dichloro-2-methylphenyl)-2-[[6-[4-(2-hydroxyethyl)piperazin-1-yl]-2-methylpyrimidin-4-yl]amino]-1,3-thiazole-5-carboxamide
SMILES OCCN(CC1)CCN1C2=NC(C)=NC(NC3=NC=C(C(NC4=C(Cl)C=CC(Cl)=C4C)=O)S3)=C2
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C22H25Cl2N7O2S/c1-13-15(23)3-4-16(24)20(13)29-21(33)17-12-25-22(34-17)28-18-11-19(27-14(2)26-18)31-7-5-30(6-8-31)9-10-32/h3-4,11-12,32H,5-10H2,1-2H3,(H,29,33)(H,25,26,27,28)
Chemical Name or Material 5-Chloro Dasatinib