Learn More
5'-Chloro-5'-deoxy-2',3'-O-isopropylideneadenosine, TRC
Specifications
Specifications
| Chemical Name or Material | 5'-Chloro-5'-deoxy-2',3'-O-isopropylideneadenosine |
| CAS | 24514-56-5 |
| Molecular Formula | C13H16ClN5O3 |
| Quantity | 100 mg |
| Synonym | 5'-Chloro-5'-deoxy-2',3'-O-(1-methylethylidene) Adenosine, Adenosine, 5′-chloro-5′-deoxy-2′,3′-O-isopropylidene- (8CI), Furo[3,4-d]-1,3-dioxole, adenosine deriv. (ZCI), 5′-Chloro-5′-deoxy-2′,3′-O-isopropylideneadenosine |
| InChI Formula | InChI=1S/C13H16ClN5O3/c1-13(2)21-8-6(3-14)20-12(9(8)22-13)19-5-18-7-10(15)16-4-17-11(7)19/h4-6,8-9,12H,3H2,1-2H3,(H2,15,16,17)/t6-,8-,9-,12-/m1/s1 |
| SMILES | C(Cl)[C@H]1O[C@H]([C@]2([C@@]1(OC(C)(C)O2)[H])[H])N3C=4C(N=C3)=C(N)N=CN4 |
| IUPAC Name | 9-((3aR,4R,6S,6aS)-6-(chloromethyl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-yl)-9H-purin-6-amine |
| Molecular Weight (g/mol) | 325.75 |
| Formula Weight | 325.09 |