missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylbenzoic acid, 98%, Thermo Scientific™

Product Code. 15457128
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15457128

Brand: Thermo Scientific Alfa Aesar H60649.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7499-06-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00045842

Specifications

Melting Point 168°C to 173°C
Quantity 1 g
Synonym 2-methyl-5-chlorobenzoic acid, 5-chloro-o-toluic acid, 5-chloro-2-methyl-benzoic acid, benzoic acid, 5-chloro-2-methyl, o-toluic acid, 5-chloro, 5-chloro-2-methylbenzoicacid, 5-chloro-2-methyl benzoic acid, 3-chloro-6-methylbenzoic acid, pubchem4578, ksc173q5r
InChI Key CSAPESWNZDOAFU-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)Cl)C(=O)O
IUPAC Name 5-chloro-2-methylbenzoic acid
PubChem CID 82009
Formula Weight 170.59
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylbenzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.