CAS RN 7499-06-1
CAS RN 7499-06-1
IUPAC Name:
5-chloro-2-methylbenzoic acid
Synonyms:
5-chloro-2-methylbenzoic acid
Molecular Weight (g/mol):
170.59
Molecular Formula:
C8H7ClO2
InChi Key:
CSAPESWNZDOAFU-UHFFFAOYSA-N
SMILES:
CC1=CC=C(Cl)C=C1C(O)=O
1
–
1
of
1
results
1
5-Chloro-2-methylbenzoic acid, 98%, Thermo Scientific™
CAS: 7499-06-1 Molecular Formula: C8H7ClO2 Molecular Weight (g/mol): 170.59 MDL Number: MFCD00045842