missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methylaniline, Sigma-Aldrich™

Product Code. 15898800
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15898800 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15898800 Supplier Sigma Aldrich Supplier No. 8026410250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 95-79-4
Melting Point 22°C
Density 1.17 g/cm3 at 20°C
Boiling Point 237°C/1013 hPa
Molecular Formula 5-(Cl)-2-(CH3)C6H3NH2
MDL Number MFCD00007779
Quantity 250 mL
Synonym 5-Chloro-2-methylaniline, 4-Chloro-2-toluidine, 5-Chloro-o-toluidine, 2-Amino-4-chlorotoluene, 5-Chloro-2-methylaniline
Molecular Weight (g/mol) 141.6
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.