missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methyl-3-isothiazolone (~90%), TRC

Product Code. 18083610
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18083610 2.5 mg 2.5mg
17949839 25 mg 25mg
18040400 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18083610 Supplier Toronto Research Chemicals Supplier No. TRCC3692352.5MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26172-55-4
Molecular Formula C4 H4 Cl N O S
Molecular Weight (g/mol) 149.6
Synonym 4-Isothiazolin-3-one, 5-chloro-2-methyl- (8CI), 5-Chloro-2-methyl-3(2H)-isothiazolone, 2-Methyl-5-chloro-3-isothiazolone, 2-Methyl-5-chloroisothiazolin-3-one, 5-Chloro-2-methyl-2H-isothiazol-3-one, 5-Chloro-2-methyl-2H-isothiazolin-3-one, 5-Chloro-2-methyl-3(2H)-isothiazolinone, 5-Chloro-2-methyl-3-isothiazolone, 5-Chloro-2-methyl-4-isothiazolin-3-one, 5-Chloro-2-methyl-4-isothiazoline-3-ketone, 5-Chloro-2-methylisothiazolin-3-one, 5-Chloro-N-methylisothiazolin-3-one, 5-Chloro-N-methylisothiazolone, A 33, A 33 (bactericide), Bioace, CIT, CMIT, HS 818, HS 818 (antiseptic), Kathon CG 5243, Kathon IXE, Methylchloroisothiazolinone, N-Methyl-5-chloroisothiazolin-3-one, N-Methyl-5-chloroisothiazolone, T 360
IUPAC Name 5-chloro-2-methyl-1,2-thiazol-3-one
SMILES CN1SC(=CC1=O)Cl

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C4H4ClNOS/c1-6-4(7)2-3(5)8-6/h2H,1H3
Formula Weight 148.9702
Percent Purity >95%
Chemical Name or Material 5-Chloro-2-methyl-4-isothiazolin-3-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.